Product Details

SNP ID
rs141729880
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18315961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGGGCCAAGTTTTCGAGGGACTTC[A/G]TTCTTATTTCCGAGTTCTCTGAGCA
Phenotype
MIM: 601243
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SMCR8 PubMed Links

Gene Details

Gene
SMCR8
Gene Name
Smith-Magenis syndrome chromosome region, candidate 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144775.2 682 Missense Mutation ATT,GTT I58V NP_658988.2
Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320759.1 682 Intron NP_001307688.1
NM_004618.4 682 Intron NP_004609.1
XM_011524001.2 682 Intron XP_011522303.1

View Full Product Details