Product Details

SNP ID
rs141839684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4006050 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTATGTGGGTTTTTACTCTTAAAAT[A/C]AGATATCCTCTGATCATCTTCACGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZZEF1 PubMed Links

Gene Details

Gene
ZZEF1
Gene Name
zinc finger ZZ-type and EF-hand domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015113.3 9854 UTR 3 NP_055928.3
XM_005256561.2 9854 UTR 3 XP_005256618.1
XM_011523759.2 9854 Intron XP_011522061.1
XM_017024382.1 9854 Intron XP_016879871.1
XM_017024383.1 9854 Intron XP_016879872.1
XM_017024384.1 9854 Intron XP_016879873.1

View Full Product Details