Product Details

SNP ID
rs142089703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28618887 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGTGCCTGAGCAAAGTAAAACTC[A/G]GTGCGACGGACCACACTCACATCTT
Phenotype
MIM: 610664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KIAA0100 PubMed Links

Gene Details

Gene
KIAA0100
Gene Name
KIAA0100
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014680.3 5830 Silent Mutation ACC,ACT T1909T NP_055495.2
XM_005258073.2 5830 Silent Mutation ACC,ACT T1908T XP_005258130.1
XM_017025423.1 5830 Silent Mutation ACC,ACT T1766T XP_016880912.1
Gene
SGK494
Gene Name
uncharacterized serine/threonine-protein kinase SgK494
There are no transcripts associated with this gene.

Gene
SPAG5-AS1
Gene Name
SPAG5 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details