Product Details

SNP ID
rs142139534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1635055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTTTTCCGATTCAGGGCCTGGCG[C/T]GGAGGCTTAGGGATGGCCCTCTTGA
Phenotype
MIM: 607873 MIM: 610791
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCARF1 PubMed Links

Gene Details

Gene
SCARF1
Gene Name
scavenger receptor class F member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003693.3 1189 Silent Mutation CCA,CCG P732P NP_003684.2
NM_145350.2 1189 UTR 3 NP_663325.1
XM_017025256.1 1189 Silent Mutation CCA,CCG P321P XP_016880745.1
Gene
SLC43A2
Gene Name
solute carrier family 43 member 2
There are no transcripts associated with this gene.

View Full Product Details