Product Details

SNP ID
rs142291012
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:74919782 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTGCCCAGGCTGTCATCGTCCA[G/T]GCTGGGGGAGCTCTGCAGCCGACCC
Phenotype
MIM: 607827 MIM: 607696
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
OTOP2 PubMed Links

Gene Details

Gene
OTOP2
Gene Name
otopetrin 2
There are no transcripts associated with this gene.

Gene
USH1G
Gene Name
USH1 protein network component sans
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282489.2 1217 Missense Mutation ATG,CTG M249L NP_001269418.1
NM_173477.4 1217 Missense Mutation ATG,CTG M352L NP_775748.2
XM_011524296.2 1217 Missense Mutation ATG,CTG M249L XP_011522598.1

View Full Product Details