Product Details

SNP ID
rs142458559
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:43012230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGACTCAAGATGAGGCTGTGG[A/G]ACCTGCAGCAGCTGAGAAAGGAGCT
Phenotype
MIM: 600735 MIM: 607526 MIM: 604631
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFI35 PubMed Links

Gene Details

Gene
IFI35
Gene Name
interferon induced protein 35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005533.4 281 Missense Mutation AAC,GAC N25D NP_005524.2
XM_005257302.4 281 Missense Mutation AAC,GAC N25D XP_005257359.1
XM_017024584.1 281 Missense Mutation AAC,GAC N5D XP_016880073.1
XM_017024585.1 281 Missense Mutation AAC,GAC N5D XP_016880074.1
Gene
RPL27
Gene Name
ribosomal protein L27
There are no transcripts associated with this gene.

Gene
VAT1
Gene Name
vesicle amine transport 1
There are no transcripts associated with this gene.

View Full Product Details