Product Details

SNP ID
rs142558586
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:9023167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCGGCGACGAGAACGAGGACGACT[C/T]GGAGCTGGCGCGCGACTCGTACTTC
Phenotype
MIM: 601614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NTN1 PubMed Links

Gene Details

Gene
NTN1
Gene Name
netrin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004822.2 1001 Missense Mutation TCG,TTG S265L NP_004813.2
XM_006721595.3 1001 Missense Mutation TCG,TTG S265L XP_006721658.1

View Full Product Details