Product Details

SNP ID
rs143521615
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8797456 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCACTCACCTCCATGGACAGCAG[C/T]TTCGAGTACTTGATTTTCCGCTCTC
Phenotype
MIM: 611462
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MFSD6L PubMed Links

Gene Details

Gene
MFSD6L
Gene Name
major facilitator superfamily domain containing 6 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152599.3 1894 Silent Mutation AAA,AAG K555K NP_689812.3
Gene
PIK3R6
Gene Name
phosphoinositide-3-kinase regulatory subunit 6
There are no transcripts associated with this gene.

View Full Product Details