Product Details

SNP ID
rs143697299
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:55267809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGATGATGAGAGTAACAGCCCGAC[A/G]GTCCCCCAGTCGGCATTCCTGGGGC
Phenotype
MIM: 142385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HLF PubMed Links

Gene Details

Gene
HLF
Gene Name
HLF, PAR bZIP transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002126.4 153 Silent Mutation ACA,ACG T58T NP_002117.1
XM_005257269.2 153 Silent Mutation ACA,ACG T58T XP_005257326.1
XM_011524705.1 153 UTR 5 XP_011523007.1
XM_017024556.1 153 UTR 5 XP_016880045.1

View Full Product Details