Product Details
- SNP ID
-
rs143699377
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:29616006 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGGCCGGAGGGCGGGCGCACGTCC[A/G]GGATGTTGCGCTTCGTGACCCGGAG
- Phenotype
-
MIM: 615124
MIM: 606779
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANKRD13B
PubMed Links
Gene Details
- Gene
- ANKRD13B
- Gene Name
- ankyrin repeat domain 13B
There are no transcripts associated with this gene.
- Gene
- CORO6
- Gene Name
- coronin 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_032854.3 |
1174 |
Missense Mutation |
CCG,CTG |
P411L |
NP_116243.2 |
XM_005258047.3 |
1174 |
Missense Mutation |
CCG,CTG |
P411L |
XP_005258104.1 |
XM_005258048.4 |
1174 |
Missense Mutation |
CCG,CTG |
P482L |
XP_005258105.2 |
XM_005258056.4 |
1174 |
Missense Mutation |
CCG,CTG |
P177L |
XP_005258113.1 |
XM_011525385.2 |
1174 |
Missense Mutation |
CCG,CTG |
P482L |
XP_011523687.1 |
XM_011525386.2 |
1174 |
Missense Mutation |
CCG,CTG |
P447L |
XP_011523688.1 |
XM_011525387.2 |
1174 |
Missense Mutation |
CCG,CTG |
P441L |
XP_011523689.1 |
XM_011525388.2 |
1174 |
Intron |
|
|
XP_011523690.1 |
XM_011525389.2 |
1174 |
Intron |
|
|
XP_011523691.1 |
XM_011525390.2 |
1174 |
Missense Mutation |
CCG,CTG |
P177L |
XP_011523692.1 |
XM_011525391.2 |
1174 |
Missense Mutation |
CCG,CTG |
P177L |
XP_011523693.1 |
XM_017025235.1 |
1174 |
Missense Mutation |
CCG,CTG |
P370L |
XP_016880724.1 |
- Gene
- SSH2
- Gene Name
- slingshot protein phosphatase 2
There are no transcripts associated with this gene.
View Full Product Details