Product Details

SNP ID
rs144501267
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39727729 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGATGTTCGGCCCCAGCCCCCTTC[A/G]CCCCGAGAGGGCCCTCTGCCTGCTG
Phenotype
MIM: 164870 MIM: 611802
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERBB2 PubMed Links

Gene Details

Gene
ERBB2
Gene Name
erb-b2 receptor tyrosine kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005862.2 3939 Silent Mutation TCA,TCG S1121S NP_001005862.1
NM_001289936.1 3939 Silent Mutation TCA,TCG S1136S NP_001276865.1
NM_001289937.1 3939 UTR 3 NP_001276866.1
NM_001289938.1 3939 Intron NP_001276867.1
NM_004448.3 3939 Silent Mutation TCA,TCG S1151S NP_004439.2
Gene
MIEN1
Gene Name
migration and invasion enhancer 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032339.4 3939 Intron NP_115715.3
XM_005257736.3 3939 Intron XP_005257793.1
Gene
MIR4728
Gene Name
microRNA 4728
There are no transcripts associated with this gene.

View Full Product Details