Product Details

SNP ID
rs144615958
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28898649 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCCCAGGCCCAAGCCCTGCCAGC[C/T]TCTTGCTGGCCTCCCATAGCCGATG
Phenotype
MIM: 616157 MIM: 131560
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144683.3 1053 Missense Mutation AAG,AGG K309R NP_653284.2
Gene
FLOT2
Gene Name
flotillin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004475.2 1053 Intron NP_004466.2
XM_005257950.3 1053 Intron XP_005258007.1
XM_005257953.2 1053 Intron XP_005258010.1
XM_017024394.1 1053 Intron XP_016879883.1
XM_017024395.1 1053 Intron XP_016879884.1
XM_017024396.1 1053 Intron XP_016879885.1
Gene
PHF12
Gene Name
PHD finger protein 12
There are no transcripts associated with this gene.

View Full Product Details