Product Details

SNP ID
rs144813252
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:47744329 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCCAAGAAACCCAGTTCATTGC[C/T]GTGACTGCCTACCAGAATGCCGAGG
Phenotype
MIM: 604895
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TBX21 PubMed Links

Gene Details

Gene
TBX21
Gene Name
T-box 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013351.1 1181 Silent Mutation GCC,GCT A301A NP_037483.1
XM_011524698.2 1181 Silent Mutation GCC,GCT A322A XP_011523000.1

View Full Product Details