Product Details

SNP ID
rs144903710
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35984395 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCATAGTAATCCATAATCCGCTGA[C/T]GCGGGATCTTGTAGGTAGTGTAGGT
Phenotype
MIM: 601392 MIM: 601394
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL14 PubMed Links

Gene Details

Gene
CCL14
Gene Name
C-C motif chemokine ligand 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032962.4 264 Missense Mutation CAT,CGT H62R NP_116738.1
NM_032963.3 264 Missense Mutation CAT,CGT H46R NP_116739.1
Gene
CCL15-CCL14
Gene Name
CCL15-CCL14 readthrough (NMD candidate)
There are no transcripts associated with this gene.

Gene
CCL16
Gene Name
C-C motif chemokine ligand 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004590.3 264 Intron NP_004581.1
XM_005258020.4 264 Intron XP_005258077.1
Gene
LOC101927339
Gene Name
uncharacterized LOC101927339
There are no transcripts associated with this gene.

View Full Product Details