Product Details

SNP ID
rs145034256
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:50114377 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACCATCCCTATCTGGGTGGAGC[C/T]GAAGTCCTGGACTAGGCACCAGGAG
Phenotype
MIM: 602525
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
PDK2 PubMed Links

Gene Details

Gene
PDK2
Gene Name
pyruvate dehydrogenase kinase 2
There are no transcripts associated with this gene.

Gene
SAMD14
Gene Name
sterile alpha motif domain containing 14
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001257359.1 2119 Intron NP_001244288.1
NM_174920.3 2119 Missense Mutation NP_777580.1
XM_011524490.2 2119 Intron XP_011522792.1
XM_017024322.1 2119 Intron XP_016879811.1
XM_017024323.1 2119 Intron XP_016879812.1
XM_017024324.1 2119 Missense Mutation XP_016879813.1

View Full Product Details