Product Details

SNP ID
rs145134017
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:7481939 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGCATCTGCCCTGCCCCAGGT[C/T]GCCTGGGCTGCGGCTGCTGTCTACC
Phenotype
MIM: 180660 MIM: 612308
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POLR2A PubMed Links

Gene Details

Gene
POLR2A
Gene Name
RNA polymerase II subunit A
There are no transcripts associated with this gene.

Gene
SLC35G6
Gene Name
solute carrier family 35 member G6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001102614.1 Intron NP_001096084.1
Gene
ZBTB4
Gene Name
zinc finger and BTB domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128833.1 Intron NP_001122305.1
NM_020899.3 Intron NP_065950.2
XM_006721563.3 Intron XP_006721626.1
XM_006721564.2 Intron XP_006721627.1
XM_011523972.2 Intron XP_011522274.1

View Full Product Details