Product Details

SNP ID
rs145468395
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48575755 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGAGAGGGCTCTGCAACATCCTC[C/T]TCCCAGACTGGGAGGGGCACATTTT
Phenotype
MIM: 142966 MIM: 142965 MIM: 610173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXB3 PubMed Links

Gene Details

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 1785 Intron NP_002137.4
XM_005257277.3 1785 Intron XP_005257334.1
XM_005257280.3 1785 Intron XP_005257337.1
XM_005257282.3 1785 Intron XP_005257339.1
XM_006721854.2 1785 Intron XP_006721917.1
XM_011524707.2 1785 Intron XP_011523009.1
XM_011524708.2 1785 Intron XP_011523010.1
XM_011524710.1 1785 Intron XP_011523012.1
XM_011524719.1 1785 Intron XP_011523021.1
XM_011524720.2 1785 Intron XP_011523022.1
XM_011524721.2 1785 Intron XP_011523023.1
XM_011524726.2 1785 Intron XP_011523028.1
XM_017024557.1 1785 Intron XP_016880046.1
XM_017024558.1 1785 Intron XP_016880047.1
XM_017024559.1 1785 Intron XP_016880048.1
XM_017024560.1 1785 Intron XP_016880049.1
XM_017024561.1 1785 Intron XP_016880050.1
XM_017024562.1 1785 Intron XP_016880051.1
XM_017024563.1 1785 Intron XP_016880052.1
Gene
HOXB4
Gene Name
homeobox B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024015.4 1785 UTR 3 NP_076920.1
Gene
MIR10A
Gene Name
microRNA 10a
There are no transcripts associated with this gene.

View Full Product Details