Product Details

SNP ID
rs145658151
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8797668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCACTCAGAGCCCTCTCCATGCGGG[C/G]AGTGGCCAGGTCCTCTACTGAGGCC
Phenotype
MIM: 611462
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MFSD6L PubMed Links

Gene Details

Gene
MFSD6L
Gene Name
major facilitator superfamily domain containing 6 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152599.3 1682 Missense Mutation CCC,GCC P485A NP_689812.3
Gene
PIK3R6
Gene Name
phosphoinositide-3-kinase regulatory subunit 6
There are no transcripts associated with this gene.

View Full Product Details