Product Details

SNP ID
rs146269888
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3197993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATTGGAAACACCAGTGCTCTCCCC[C/T]ACACTCTGCTCACAGCTAGTTTGTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR1A2 PubMed Links

Gene Details

Gene
OR1A2
Gene Name
olfactory receptor family 1 subfamily A member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012352.2 477 Missense Mutation CAC,TAC H159Y NP_036484.1

View Full Product Details