Product Details

SNP ID
rs146410774
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:10494935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGCTGATTAGGAGACCCACCCTG[C/G]CCTCCAGTTTCTGGATCTGCTTCTT
Phenotype
MIM: 160730
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MYH1 PubMed Links

Gene Details

Gene
MYH1
Gene Name
myosin heavy chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005963.3 5729 Missense Mutation GCC,GGC A1821G NP_005954.3
XM_017024675.1 5729 Missense Mutation GCC,GGC A1821G XP_016880164.1
Gene
MYHAS
Gene Name
myosin heavy chain gene cluster antisense RNA
There are no transcripts associated with this gene.

View Full Product Details