Product Details

SNP ID
rs146651013
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28898753 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATATCTCCCACTGAGGGGCTC[A/G]ATGCCCTCTTGTAGAGCACAATACA
Phenotype
MIM: 616157 MIM: 131560
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144683.3 949 Silent Mutation ATC,ATT I274I NP_653284.2
Gene
FLOT2
Gene Name
flotillin 2
There are no transcripts associated with this gene.

Gene
PHF12
Gene Name
PHD finger protein 12
There are no transcripts associated with this gene.

View Full Product Details