Product Details
- SNP ID
-
rs146811066
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:17248962 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGTTATGAAGCATGGCTGGGTTATT[A/G]TATTTTTCAGGGTTATCATGGAAAC
- Phenotype
-
MIM: 604665
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COPS3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs553573176] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COPS3
- Gene Name
- COP9 signalosome subunit 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001199125.1 |
1076 |
Silent Mutation |
TAC,TAT |
Y347Y |
NP_001186054.1 |
NM_001316354.1 |
1076 |
Silent Mutation |
TAC,TAT |
Y237Y |
NP_001303283.1 |
NM_001316355.1 |
1076 |
Silent Mutation |
TAC,TAT |
Y309Y |
NP_001303284.1 |
NM_001316356.1 |
1076 |
Silent Mutation |
TAC,TAT |
Y300Y |
NP_001303285.1 |
NM_001316357.1 |
1076 |
Silent Mutation |
TAC,TAT |
Y262Y |
NP_001303286.1 |
NM_001316358.1 |
1076 |
Silent Mutation |
TAC,TAT |
Y262Y |
NP_001303287.1 |
NM_003653.3 |
1076 |
Silent Mutation |
TAC,TAT |
Y367Y |
NP_003644.2 |
XM_005256837.4 |
1076 |
Silent Mutation |
TAC,TAT |
Y347Y |
XP_005256894.1 |
XM_005256840.3 |
1076 |
Silent Mutation |
TAC,TAT |
Y237Y |
XP_005256897.1 |
XM_005256842.4 |
1076 |
Silent Mutation |
TAC,TAT |
Y237Y |
XP_005256899.1 |
XM_017025246.1 |
1076 |
Silent Mutation |
TAC,TAT |
Y262Y |
XP_016880735.1 |
View Full Product Details