Product Details

SNP ID
rs147038217
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59159900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTATTTCTCCATTAATATCATA[C/T]ACATATCCCATCATAACCCATACAC
Phenotype
MIM: 615675 MIM: 615920 MIM: 616674
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR301A PubMed Links

Gene Details

Gene
MIR301A
Gene Name
microRNA 301a
There are no transcripts associated with this gene.

Gene
PRR11
Gene Name
proline rich 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018304.3 Intron NP_060774.2
Gene
SKA2
Gene Name
spindle and kinetochore associated complex subunit 2
There are no transcripts associated with this gene.

View Full Product Details