Product Details

SNP ID
rs147208105
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:40389125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTATGCTCTATCTCATATCTACT[A/G]AAAGTAGAAAAGGAGGAAGAGTGAC
Phenotype
MIM: 126430
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOP2A PubMed Links

Gene Details

Gene
TOP2A
Gene Name
topoisomerase (DNA) II alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001067.3 5091 UTR 3 NP_001058.2
XM_005257632.1 5091 UTR 3 XP_005257689.1
XM_011525165.1 5091 Intron XP_011523467.1

View Full Product Details