Product Details
- SNP ID
-
rs147400711
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:42568554 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCACCAGTGCCTCTTCTGTCCCCA[A/G]CACAGGTAGGCAGTAACATCCCCCC
- Phenotype
-
MIM: 609855
MIM: 602976
MIM: 608665
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
COASY
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs646123] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- COASY
- Gene Name
- Coenzyme A synthase
There are no transcripts associated with this gene.
- Gene
- MLX
- Gene Name
- MLX, MAX dimerization protein
- Gene
- PSMC3IP
- Gene Name
- PSMC3 interacting protein
There are no transcripts associated with this gene.
View Full Product Details