Product Details

SNP ID
rs147458712
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:28901179 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGCAGCTGAGGCTACCACCACCA[C/T]GCGGCTAGGGGCACATGCCTTCAGG
Phenotype
MIM: 616157 MIM: 131560
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
DHRS13 PubMed Links
Additional Information
For this assay, SNP(s) [rs4795474] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
DHRS13
Gene Name
dehydrogenase/reductase 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144683.3 620 Missense Mutation NP_653284.2
Gene
FLOT2
Gene Name
flotillin 2
There are no transcripts associated with this gene.

Gene
PHF12
Gene Name
PHD finger protein 12
There are no transcripts associated with this gene.

View Full Product Details