Product Details

SNP ID
rs147489840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8072868 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCGGATGTCGTGTGAGATCTGG[C/T]TCAGGCGCTGGCGGAACGCCTCTAT
Phenotype
MIM: 603741
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALOX12B PubMed Links

Gene Details

Gene
ALOX12B
Gene Name
arachidonate 12-lipoxygenase, 12R type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001139.2 2270 Missense Mutation AAC,AGC N670S NP_001130.1
Gene
LOC105371518
Gene Name
uncharacterized LOC105371518
There are no transcripts associated with this gene.

View Full Product Details