Product Details

SNP ID
rs147829173
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:35872389 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGACTCTCCATCCTAGCTCATCTC[C/T]AAAGAGTTGATGTACTCCCGAACCC
Phenotype
MIM: 187011
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCL5 PubMed Links

Gene Details

Gene
CCL5
Gene Name
C-C motif chemokine ligand 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278736.1 332 Missense Mutation AGA,GGA R116G NP_001265665.1
NM_002985.2 332 Silent Mutation TTA,TTG L88L NP_002976.2
Gene
HEATR9
Gene Name
HEAT repeat containing 9
There are no transcripts associated with this gene.

View Full Product Details