Product Details

SNP ID
rs148027903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:15651290 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGTTTGATGTTCACCTGATGC[A/G]GCTGCAAGTGCTCTTCACAGTAATT
Phenotype
MIM: 609505
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRIM16 PubMed Links

Gene Details

Gene
TRIM16
Gene Name
tripartite motif containing 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006470.3 877 Missense Mutation NP_006461.3

View Full Product Details