Product Details
- SNP ID
-
rs148146423
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:9897959 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGTGCTGTGGGCTTGTGTGCAGG[C/T]CTTTCTCTTGGCCCTGGGGTGGATG
- Phenotype
-
MIM: 603659
MIM: 179618
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GLP2R
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs531533066] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLP2R
- Gene Name
- glucagon like peptide 2 receptor
There are no transcripts associated with this gene.
- Gene
- RCVRN
- Gene Name
- recoverin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002903.2 |
926 |
UTR 3 |
|
|
NP_002894.1 |
View Full Product Details