Product Details

SNP ID
rs148289755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81922802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTCGAGCTCCAGCTTCATGCTGG[C/T]GTTCTCTGAGGCCAGCTTCTCCACC
Phenotype
MIM: 602020 MIM: 179035 MIM: 606212
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAFG PubMed Links

Gene Details

Gene
MAFG
Gene Name
MAF bZIP transcription factor G
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002359.3 505 Missense Mutation ACC,GCC T98A NP_002350.1
NM_032711.3 505 Missense Mutation ACC,GCC T98A NP_116100.2
XM_011523578.1 505 Missense Mutation ACC,GCC T98A XP_011521880.1
Gene
MAFG-AS1
Gene Name
MAFG antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

Gene
PYCR1
Gene Name
pyrroline-5-carboxylate reductase 1
There are no transcripts associated with this gene.

Gene
SIRT7
Gene Name
sirtuin 7
There are no transcripts associated with this gene.

View Full Product Details