Product Details

SNP ID
rs148677602
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39260110 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATTAACACTGACATACAATTGA[C/G]CAGCTGTTATAGAGCCATTCTTGTA
Phenotype
MIM: 609086
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FBXL20 PubMed Links

Gene Details

Gene
FBXL20
Gene Name
F-box and leucine rich repeat protein 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184906.1 2915 UTR 3 NP_001171835.1
NM_032875.2 2915 UTR 3 NP_116264.2
XM_005257746.3 2915 UTR 3 XP_005257803.2
XM_005257747.3 2915 UTR 3 XP_005257804.2

View Full Product Details