Product Details
- SNP ID
-
rs148682580
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40792507 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAATGGAGTGAATCCTTGATGAAA[G/T]AACTTTACCACGTTGATCTAGCTCT
- Phenotype
-
MIM: 616676
MIM: 616677
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT27
PubMed Links
Gene Details
- Gene
- KRT27
- Gene Name
- keratin 27
There are no transcripts associated with this gene.
- Gene
- KRT28
- Gene Name
- keratin 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181535.3 |
1381 |
Missense Mutation |
ATT,CTT |
I439L |
NP_853513.2 |
View Full Product Details