Product Details

SNP ID
rs148715809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8204933 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCGAGAGTTGGCCCGGACCCAAG[A/G]GTGGGCTGAGACCTGGGCCAGGGGC
Phenotype
MIM: 604970
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AURKB PubMed Links

Gene Details

Gene
AURKB
Gene Name
aurora kinase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256834.2 1086 Missense Mutation NP_001243763.1
NM_001284526.1 1086 Missense Mutation NP_001271455.1
NM_001313950.1 1086 Missense Mutation NP_001300879.1
NM_001313951.1 1086 Intron NP_001300880.1
NM_001313952.1 1086 Missense Mutation NP_001300881.1
NM_001313953.1 1086 Missense Mutation NP_001300882.1
NM_001313954.1 1086 Missense Mutation NP_001300883.1
NM_001313955.1 1086 Missense Mutation NP_001300884.1
NM_004217.3 1086 Missense Mutation NP_004208.2
XM_011524070.2 1086 Missense Mutation XP_011522372.1
XM_011524072.2 1086 Missense Mutation XP_011522374.1
XM_017025307.1 1086 Missense Mutation XP_016880796.1
XM_017025308.1 1086 Missense Mutation XP_016880797.1
XM_017025309.1 1086 Missense Mutation XP_016880798.1
XM_017025310.1 1086 Missense Mutation XP_016880799.1
XM_017025311.1 1086 Missense Mutation XP_016880800.1

View Full Product Details