Product Details

SNP ID
rs148915379
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48623125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGCGAGCGTGCAGCCAGTTGGC[A/G]GAGGGGTTGGCTGAAAGAGAAGCAG
Phenotype
MIM: 142963 MIM: 142964 MIM: 608632
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HOXB8 PubMed Links

Gene Details

Gene
HOXB8
Gene Name
homeobox B8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024016.3 732 Intron NP_076921.1
XM_005257286.2 732 Intron XP_005257343.1
XM_017024564.1 732 Intron XP_016880053.1
Gene
HOXB9
Gene Name
homeobox B9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024017.4 732 Silent Mutation TCC,TCT S176S NP_076922.1
Gene
MIR196A1
Gene Name
microRNA 196a-1
There are no transcripts associated with this gene.

View Full Product Details