Product Details

SNP ID
rs149343575
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:59708905 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGTATGAAGTGACTCTCTCGAGTA[G/T]AAATTTTATATTTTACTACTTTGCC
Phenotype
MIM: 608625 MIM: 611753
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PTRH2 PubMed Links

Gene Details

Gene
PTRH2
Gene Name
peptidyl-tRNA hydrolase 2
There are no transcripts associated with this gene.

Gene
VMP1
Gene Name
vacuole membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details