Product Details

SNP ID
rs149849406
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:18315802 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATATCTGGAAATATGATCAGCGCC[A/C]CTGACGTAGTGGCCTTCACCAAAGA
Phenotype
MIM: 601243
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SMCR8 PubMed Links

Gene Details

Gene
SMCR8
Gene Name
Smith-Magenis syndrome chromosome region, candidate 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144775.2 523 Missense Mutation ACT,CCT T5P NP_658988.2
Gene
TOP3A
Gene Name
topoisomerase (DNA) III alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320759.1 523 Intron NP_001307688.1
NM_004618.4 523 Intron NP_004609.1
XM_011524001.2 523 Intron XP_011522303.1

View Full Product Details