Product Details
- SNP ID
-
rs150448547
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40793988 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCCCCGATCTGAGCCTGGATCTGC[A/G]CCAGCTGCGTACAGTAGTTGCTCTC
- Phenotype
-
MIM: 616677
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT28
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2270153] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT28
- Gene Name
- keratin 28
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_181535.3 |
1103 |
Missense Mutation |
GCG,GTG |
A346V |
NP_853513.2 |
View Full Product Details