Product Details

SNP ID
rs150754782
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4790089 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGACGCCTACCGTGCGGCCCTGGG[C/T]CCGCATCACCCCTGGCTGGTCCGAC
Phenotype
MIM: 600307 MIM: 604657
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLTPD2 PubMed Links

Gene Details

Gene
GLTPD2
Gene Name
glycolipid transfer protein domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014985.2 873 Silent Mutation GGC,GGT G223G NP_001014985.2
XM_005256626.3 873 Silent Mutation GGC,GGT G259G XP_005256683.1
Gene
PSMB6
Gene Name
proteasome subunit beta 6
There are no transcripts associated with this gene.

Gene
TM4SF5
Gene Name
transmembrane 4 L six family member 5
There are no transcripts associated with this gene.

Gene
VMO1
Gene Name
vitelline membrane outer layer 1 homolog
There are no transcripts associated with this gene.

View Full Product Details