Product Details

SNP ID
rs150798403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:8226811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCGGTTGCACGTGAACCTTCCT[C/T]TCACCCGGGTGCCCAGAGACTCCTC
Phenotype
MIM: 613129
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTC1 PubMed Links

Gene Details

Gene
CTC1
Gene Name
CTS telomere maintenance complex component 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025099.5 4831 UTR 3 NP_079375.3
XM_006721577.3 4831 UTR 3 XP_006721640.1
XM_006721578.3 4831 UTR 3 XP_006721641.1
XM_011524010.2 4831 UTR 3 XP_011522312.1
XM_011524011.2 4831 UTR 3 XP_011522313.1
Gene
LINC00324
Gene Name
long intergenic non-protein coding RNA 324
There are no transcripts associated with this gene.

View Full Product Details