Product Details
- SNP ID
-
rs139856475
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:37253827 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CGCCGGCGCTCAGTACGGGCGATTG[A/G]CGTCTTTGGGCCGCTTCAGCTGCAC
- Phenotype
-
MIM: 612679
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CELF4
PubMed Links
Gene Details
- Gene
- CELF4
- Gene Name
- CUGBP, Elav-like family member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001025087.1 |
1817 |
Missense Mutation |
GCC,GTC |
A481V |
NP_001020258.1 |
NM_001025088.1 |
1817 |
Missense Mutation |
GCC,GTC |
A480V |
NP_001020259.1 |
NM_001025089.1 |
1817 |
Missense Mutation |
GCC,GTC |
A444V |
NP_001020260.1 |
NM_020180.3 |
1817 |
Missense Mutation |
GCC,GTC |
A482V |
NP_064565.1 |
XM_005258308.3 |
1817 |
Missense Mutation |
GCC,GTC |
A480V |
XP_005258365.1 |
XM_011526079.2 |
1817 |
Missense Mutation |
GCC,GTC |
A502V |
XP_011524381.1 |
XM_011526081.2 |
1817 |
Missense Mutation |
GCC,GTC |
A501V |
XP_011524383.1 |
XM_011526083.2 |
1817 |
Missense Mutation |
GCC,GTC |
A501V |
XP_011524385.1 |
XM_011526084.2 |
1817 |
Missense Mutation |
GCC,GTC |
A500V |
XP_011524386.1 |
XM_011526085.2 |
1817 |
Missense Mutation |
GCC,GTC |
A500V |
XP_011524387.1 |
XM_011526086.2 |
1817 |
Missense Mutation |
GCC,GTC |
A491V |
XP_011524388.1 |
XM_011526087.2 |
1817 |
Missense Mutation |
GCC,GTC |
A490V |
XP_011524389.1 |
XM_011526090.2 |
1817 |
Missense Mutation |
GCC,GTC |
A481V |
XP_011524392.1 |
XM_011526091.2 |
1817 |
Missense Mutation |
GCC,GTC |
A454V |
XP_011524393.1 |
XM_011526092.2 |
1817 |
Missense Mutation |
GCC,GTC |
A453V |
XP_011524394.1 |
XM_017025840.1 |
1817 |
Missense Mutation |
GCC,GTC |
A492V |
XP_016881329.1 |
XM_017025841.1 |
1817 |
Missense Mutation |
GCC,GTC |
A481V |
XP_016881330.1 |
XM_017025842.1 |
1817 |
Missense Mutation |
GCC,GTC |
A481V |
XP_016881331.1 |
XM_017025843.1 |
1817 |
Missense Mutation |
GCC,GTC |
A480V |
XP_016881332.1 |
XM_017025844.1 |
1817 |
Missense Mutation |
GCC,GTC |
A480V |
XP_016881333.1 |
XM_017025845.1 |
1817 |
Missense Mutation |
GCC,GTC |
A479V |
XP_016881334.1 |
XM_017025846.1 |
1817 |
Missense Mutation |
GCC,GTC |
A472V |
XP_016881335.1 |
XM_017025847.1 |
1817 |
Missense Mutation |
GCC,GTC |
A471V |
XP_016881336.1 |
XM_017025848.1 |
1817 |
Missense Mutation |
GCC,GTC |
A470V |
XP_016881337.1 |
XM_017025849.1 |
1817 |
Missense Mutation |
GCC,GTC |
A469V |
XP_016881338.1 |
XM_017025850.1 |
1817 |
Missense Mutation |
GCC,GTC |
A471V |
XP_016881339.1 |
XM_017025851.1 |
1817 |
Missense Mutation |
GCC,GTC |
A470V |
XP_016881340.1 |
XM_017025852.1 |
1817 |
Missense Mutation |
GCC,GTC |
A470V |
XP_016881341.1 |
XM_017025853.1 |
1817 |
Missense Mutation |
GCC,GTC |
A453V |
XP_016881342.1 |
XM_017025854.1 |
1817 |
Missense Mutation |
GCC,GTC |
A453V |
XP_016881343.1 |
XM_017025855.1 |
1817 |
Missense Mutation |
GCC,GTC |
A452V |
XP_016881344.1 |
XM_017025856.1 |
1817 |
Missense Mutation |
GCC,GTC |
A452V |
XP_016881345.1 |
XM_017025857.1 |
1817 |
Intron |
|
|
XP_016881346.1 |
XM_017025858.1 |
1817 |
Missense Mutation |
GCC,GTC |
A452V |
XP_016881347.1 |
XM_017025859.1 |
1817 |
Missense Mutation |
GCC,GTC |
A451V |
XP_016881348.1 |
XM_017025860.1 |
1817 |
Missense Mutation |
GCC,GTC |
A443V |
XP_016881349.1 |
XM_017025861.1 |
1817 |
Missense Mutation |
GCC,GTC |
A443V |
XP_016881350.1 |
XM_017025862.1 |
1817 |
Missense Mutation |
GCC,GTC |
A443V |
XP_016881351.1 |
XM_017025863.1 |
1817 |
Missense Mutation |
GCC,GTC |
A442V |
XP_016881352.1 |
XM_017025864.1 |
1817 |
Missense Mutation |
GCC,GTC |
A442V |
XP_016881353.1 |
XM_017025865.1 |
1817 |
Missense Mutation |
GCC,GTC |
A442V |
XP_016881354.1 |
XM_017025866.1 |
1817 |
Missense Mutation |
GCC,GTC |
A358V |
XP_016881355.1 |
XM_017025867.1 |
1817 |
Missense Mutation |
GCC,GTC |
A357V |
XP_016881356.1 |
XM_017025868.1 |
1817 |
Missense Mutation |
GCC,GTC |
A349V |
XP_016881357.1 |
XM_017025869.1 |
1817 |
Missense Mutation |
GCC,GTC |
A348V |
XP_016881358.1 |
XM_017025870.1 |
1817 |
Missense Mutation |
GCC,GTC |
A348V |
XP_016881359.1 |
XM_017025871.1 |
1817 |
Missense Mutation |
GCC,GTC |
A321V |
XP_016881360.1 |
XM_017025872.1 |
1817 |
Missense Mutation |
GCC,GTC |
A320V |
XP_016881361.1 |
View Full Product Details