Product Details

SNP ID
rs140788289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:63328189 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTTTATGTAATTTAGCAAAAATA[G/T]ATATATAATTGCTTTCTTCTAGAAA
Phenotype
MIM: 151430 MIM: 136440
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
BCL2 PubMed Links

Gene Details

Gene
BCL2
Gene Name
BCL2, apoptosis regulator
There are no transcripts associated with this gene.

Gene
KDSR
Gene Name
3-ketodihydrosphingosine reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002035.2 4612 UTR 3 NP_002026.1
XM_005266677.2 4612 UTR 3 XP_005266734.1
XM_017025690.1 4612 UTR 3 XP_016881179.1

View Full Product Details