Product Details

SNP ID
rs141882281
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:12308746 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCGGCCGGAGGCTACGTGGGAGA[C/T]TCGGCGCTGCAGCTGGAGAGAATCA
Phenotype
MIM: 615103
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TUBB6 PubMed Links

Gene Details

Gene
TUBB6
Gene Name
tubulin beta 6 class V
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303524.1 351 Silent Mutation GAC,GAT D39D NP_001290453.1
NM_001303525.1 351 Silent Mutation GAC,GAT D39D NP_001290454.1
NM_001303526.1 351 Silent Mutation GAC,GAT D39D NP_001290455.1
NM_001303527.1 351 Intron NP_001290456.1
NM_001303528.1 351 UTR 5 NP_001290457.1
NM_001303529.1 351 UTR 5 NP_001290458.1
NM_001303530.1 351 UTR 5 NP_001290459.1
NM_032525.2 351 Silent Mutation GAC,GAT D39D NP_115914.1

View Full Product Details