Product Details
- SNP ID
-
rs143598035
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:12674674 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTGCCTGCTGATATACTTGTTGTA[C/T]GCTCAAATTCATAAGAAGCCAAAGC
- Phenotype
-
MIM: 609702
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CEP76
PubMed Links
Gene Details
- Gene
- CEP76
- Gene Name
- centrosomal protein 76
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001271989.1 |
1636 |
Missense Mutation |
CAT,CGT |
H493R |
NP_001258918.1 |
NM_024899.3 |
1636 |
Missense Mutation |
CAT,CGT |
H568R |
NP_079175.2 |
XM_005258149.4 |
1636 |
Missense Mutation |
CAT,CGT |
H568R |
XP_005258206.1 |
XM_017025981.1 |
1636 |
Missense Mutation |
CAT,CGT |
H568R |
XP_016881470.1 |
XM_017025982.1 |
1636 |
Missense Mutation |
CAT,CGT |
H547R |
XP_016881471.1 |
XM_017025983.1 |
1636 |
Missense Mutation |
CAT,CGT |
H525R |
XP_016881472.1 |
XM_017025984.1 |
1636 |
Missense Mutation |
CAT,CGT |
H390R |
XP_016881473.1 |
XM_017025985.1 |
1636 |
Missense Mutation |
CAT,CGT |
H390R |
XP_016881474.1 |
XM_017025986.1 |
1636 |
Missense Mutation |
CAT,CGT |
H390R |
XP_016881475.1 |
XM_017025987.1 |
1636 |
Missense Mutation |
CAT,CGT |
H390R |
XP_016881476.1 |
- Gene
- PSMG2
- Gene Name
- proteasome assembly chaperone 2
View Full Product Details