Product Details

SNP ID
rs144660620
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.18:76980470 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATCCAGAGCGACTATCTCTTCCT[C/T]CCTGAAAAGGAAGAGAGAGGAGTTA
Phenotype
MIM: 159430 MIM: 604760
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MBP PubMed Links

Gene Details

Gene
MBP
Gene Name
myelin basic protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001025081.1 609 Missense Mutation GAA,GGA E184G NP_001020252.1
NM_001025090.1 609 Missense Mutation GAA,GGA E158G NP_001020261.1
NM_001025092.1 609 Missense Mutation GAA,GGA E147G NP_001020263.1
NM_001025100.1 609 Intron NP_001020271.1
NM_001025101.1 609 Missense Mutation GAA,GGA E291G NP_001020272.1
NM_002385.2 609 Missense Mutation GAA,GGA E173G NP_002376.1
XM_011526009.2 609 Intron XP_011524311.1
XM_017025778.1 609 Intron XP_016881267.1
XM_017025779.1 609 Intron XP_016881268.1
XM_017025780.1 609 Intron XP_016881269.1
Gene
ZNF236
Gene Name
zinc finger protein 236
There are no transcripts associated with this gene.

View Full Product Details