Product Details

SNP ID
rs146394838
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:74148832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATGATTTGTACTTTTCTACGAGC[C/T]GTACAGTATACGGAGAAGCTGCACA
Phenotype
MIM: 609093 MIM: 615180
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
FBXO15 PubMed Links

Gene Details

Gene
FBXO15
Gene Name
F-box protein 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142958.1 306 Intron NP_001136430.1
NM_152676.2 306 Intron NP_689889.1
XM_011525856.1 306 Intron XP_011524158.1
XM_011525858.2 306 Intron XP_011524160.1
XM_011525859.1 306 Intron XP_011524161.1
XM_011525860.1 306 Intron XP_011524162.1
XM_017025603.1 306 Intron XP_016881092.1
XM_017025604.1 306 Intron XP_016881093.1
Gene
TIMM21
Gene Name
translocase of inner mitochondrial membrane 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014177.2 306 Silent Mutation GCC,GCT A8A NP_054896.2
XM_005266689.3 306 UTR 5 XP_005266746.1

View Full Product Details