Product Details

SNP ID
rs137873305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:13100667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAACCCACACTCACCCTCAGCCAG[A/G]TCCAGCTCACAGTGGCTTGGTCTCC
Phenotype
MIM: 151440 MIM: 164005 MIM: 611669
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LYL1 PubMed Links

Gene Details

Gene
LYL1
Gene Name
LYL1, basic helix-loop-helix family member
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005583.4 1624 Silent Mutation GAC,GAT D139D NP_005574.2
XM_005259912.3 1624 Silent Mutation GAC,GAT D65D XP_005259969.1
XM_017026816.1 1624 Silent Mutation GAC,GAT D210D XP_016882305.1
XM_017026817.1 1624 Silent Mutation GAC,GAT D161D XP_016882306.1
Gene
NFIX
Gene Name
nuclear factor I X
There are no transcripts associated with this gene.

Gene
TRMT1
Gene Name
tRNA methyltransferase 1
There are no transcripts associated with this gene.

View Full Product Details