Product Details

SNP ID
rs138427304
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43511564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGGCTGGCCCTGGTCTCCAA[A/C]GCCTGGCAGGGGTGGAAGAGTACAG
Phenotype
MIM: 608451
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ETHE1 PubMed Links

Gene Details

Gene
ETHE1
Gene Name
ETHE1, persulfide dioxygenase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001320867.1 163 Silent Mutation NP_001307796.1
NM_001320868.1 163 Silent Mutation NP_001307797.1
NM_001320869.1 163 Silent Mutation NP_001307798.1
NM_014297.4 163 Silent Mutation NP_055112.2
XM_005258687.3 163 Silent Mutation XP_005258744.1
Gene
PHLDB3
Gene Name
pleckstrin homology like domain family B member 3
There are no transcripts associated with this gene.

View Full Product Details