Product Details

SNP ID
rs138534991
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:12876078 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCGGCTGGAAGGCTTTCCAGAGG[G/T]CTGGCAGCTGGGCACACAGTGTGCC
Phenotype
MIM: 126350 MIM: 600599 MIM: 612256
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DNASE2 PubMed Links

Gene Details

Gene
DNASE2
Gene Name
deoxyribonuclease II, lysosomal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001375.2 1141 Missense Mutation GAC,GCC D332A NP_001366.1
Gene
KLF1
Gene Name
Kruppel like factor 1
There are no transcripts associated with this gene.

Gene
MAST1
Gene Name
microtubule associated serine/threonine kinase 1
There are no transcripts associated with this gene.

View Full Product Details