Product Details

SNP ID
rs138562401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:48297234 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCACTGGTGGAGCCGAAGGTCAC[A/G]TGGCCAGTGTTGGGGTCACCGTGCG
Phenotype
MIM: 615038 MIM: 603996
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC114 PubMed Links

Gene Details

Gene
CCDC114
Gene Name
coiled-coil domain containing 114
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144577.3 1293 Silent Mutation CAC,CAT H585H NP_653178.3
XM_005259413.3 1293 Silent Mutation CAC,CAT H622H XP_005259470.1
XM_005259414.3 1293 Silent Mutation CAC,CAT H622H XP_005259471.1
XM_005259415.3 1293 Silent Mutation CAC,CAT H622H XP_005259472.1
XM_005259416.4 1293 Silent Mutation CAC,CAT H394H XP_005259473.1
XM_011527515.2 1293 Silent Mutation CAC,CAT H585H XP_011525817.1
XM_011527516.2 1293 Silent Mutation CAC,CAT H585H XP_011525818.1
XM_011527517.1 1293 Intron XP_011525819.1
XM_017027483.1 1293 Silent Mutation CAC,CAT H530H XP_016882972.1
Gene
ZNF114
Gene Name
zinc finger protein 114
There are no transcripts associated with this gene.

View Full Product Details